Anti-CD81 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5A6]

Supplier: Biolegend
349509 349510
10758-910EA 120.5 USD
10758-910 10758-912
Anti-CD81 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 5A6]
Antibodies
APC anti-human CD81 (TAPA-1) [5A6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: 5A6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR