Anti-CD64 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 10.1]

Supplier: Biolegend
305031 305032
10757-884EA 179 USD
10757-884 10757-886
Anti-CD64 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 10.1]
Antibodies
PE/Dazzle™ 594 anti-human CD64 [10.1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Rhesus, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD64
Clonality: Monoclonal
Clone: 10.1
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Rhesus;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR