Anti-CD307 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 509f6]

Supplier: Biolegend
340305 340306
10758-628EA 167.8 USD
10758-628 10758-630
Anti-CD307 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 509f6]
Antibodies
APC anti-human CD307e (FcRL5) [509f6]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD307
Clonality: Monoclonal
Clone: 509f6
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR