Anti-CD304 Mouse Monoclonal Antibody [clone: 14H4]

Supplier: BioLegend
652302
652302-BLEA 359.19 USD
652302-BL
Anti-CD304 Mouse Monoclonal Antibody [clone: 14H4]
Antibodies
Purified anti-human CD304 [14H4]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: WB; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. Western blotting, suggested working dilution(s): Use 10 μl per 5 ml antibody dilution buffer for each mini-gel (1 μg/ml). It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD304
Clonality: Monoclonal
Clone: 14H4
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR