Anti-CD200R3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: Ba13]

Supplier: Biolegend
142207 142208
10757-264EA 147.5 USD
10757-264 10757-266
Anti-CD200R3 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: Ba13]
Antibodies
APC anti-mouse CD200R3 [Ba13]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD200R3
Clonality: Monoclonal
Clone: Ba13
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR