Sodium chloride 0.9%, Viaflex for injection
Supplier: BAXTER IV SOLUTIONS
Synonyms:
Halite
, Sea salt
, NaCl
, Saline
2B1309
68000-327CS
1670
USD
68000-327
Sodium chloride 0.9%, Viaflex for injection
Sodium chloride
- NDC number: 0338-0049-38
Formula:
NaCl MW: 58.44 g/mol |
MDL Number:
MFCD00003477 CAS Number: 7647-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...