Lubricating Oil, GAST
Supplier: Gast Manufacturing
AD220
54908-024CS 762.89 USD
54908-024
Lubricating Oil, GAST
Lubricants
Pneumatic lubricating oil mainly used for the air compressors, vacuum pumps and compressed air motors.