Forceps, 4" (10 cm) Long, Serrated Curved, 0.5 mm Tip, Roboz
Supplier: Roboz
RS-5101
100189-764EA 226.15 USD
100189-764
Forceps, 4" (10 cm) Long, Serrated Curved, 0.5 mm Tip, Roboz
Forceps
Foerster Forceps; Curved; Serrated Tips; 0.5 mm Tip Width; 4" Length
The unique characteristic of the Foerster Forceps is the octagonal handle. The unique handle is designed to provide more "feel" and control at the tip. This handle also provides a better grip while wearing gloves. The tips are very fine and serrated and provide a sure, steady grip on tissue without trauma. The Foerester forceps are excellent for fine dissection of mice and dissection under a microscope.