Anti-CD16 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 3G8]

Supplier: Biolegend
302039 302040
10757-682EA 254.23 USD
10757-682 10757-684
Anti-CD16 Mouse Monoclonal Antibody (Brilliant Violet® 605) [clone: 3G8]
Antibodies
Brilliant Violet 605™ anti-human CD16 [3G8]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Capuchin Monkey, Chimpanzee, Cynomolgus, Marmoset, Pigtailed Macaque, Rhesus, Sooty Mangabey, Squirrel Monkey; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: 3G8
Conjugation: Brilliant Violet® 605
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Capuchin Monkey;Chimpanzee;Cynomolgus;Marmoset;Pigtailed Macaque;Rhesus;Sooty Mangabey;Squirrel Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR