Anti-CD152 Mouse Monoclonal Antibody (Biotin) [clone: L3D10]

Supplier: Biolegend
349911 349912
10758-922EA 198.1 USD
10758-922 10758-924
Anti-CD152 Mouse Monoclonal Antibody (Biotin) [clone: L3D10]
Antibodies
Biotin anti-human CD152 (CTLA-4) [L3D10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ELISA Detection; Size: 500 μg

Each lot of this antibody is quality control tested by ELISA assay. For ELISA detection applications, a concentration range of 0.5 - 2.0 μg/mL is recommended. To obtain a linear standard curve, serial dilutions of human CTLA-4 recombinant protein ranging from 2500 to 39 pg/mL are recommended for each ELISA plate. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD152
Clonality: Monoclonal
Clone: L3D10
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR