Anti-CD144 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: BV13]

Supplier: Biolegend
138015 138016
10756-452EA 179 USD
10756-452 10757-182
Anti-CD144 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: BV13]
Antibodies
PE/Cy7 anti-mouse CD144 (VE-cadherin) [BV13]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD144
Clonality: Monoclonal
Clone: BV13
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR