Anti-CD11b (activated) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: CBRM1/5]

Supplier: Biolegend
301411 301412
10757-634EA 192.5 USD
10757-634 10757-636
Anti-CD11b (activated) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: CBRM1/5]
Antibodies
PE/Cy7 anti-human CD11b (activated) [CBRM1/5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD11b (activated)
Clonality: Monoclonal
Clone: CBRM1/5
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR