Anti-CD103 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: Ber-ACT8]

Supplier: Biolegend
350213 350214
10758-946EA 210.73 USD
10758-946 10758-948
Anti-CD103 Mouse Monoclonal Antibody (Brilliant Violet® 421) [clone: Ber-ACT8]
Antibodies
Brilliant Violet 421™ anti-human CD103 (Integrin αE) [Ber-ACT8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: Ber-ACT8
Conjugation: Brilliant Violet® 421
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;Cross-Reactivity: Cynomolgus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR