Anti-Asialo GM1 Rabbit Polyclonal Antibody (Alexa Fluor® 647) [clone: Poly21460]

Supplier: Biolegend
146003 146004
10756-510EA 192.5 USD
10756-510 10756-512
Anti-Asialo GM1 Rabbit Polyclonal Antibody (Alexa Fluor® 647) [clone: Poly21460]
Antibodies
Alexa Fluor® 647 anti-Asialo-GM1 [Poly21460]; Isotype: Rabbit Polyclonal IgG; Reactivity: Mouse, Rat; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Asialo GM1
Clonality: Polyclonal
Clone: Poly21460
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: Rabbit Polyclonal IgG
Reactivity: Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR