Viva Biphenyl Columns, Restek

Supplier: Restek
9516511 9516512 9516515 9516522 9516525 9516532 9516535 9516551 9516552 9516555 9516561 9516562 9516565 951651E 951652E 951653E 951655E 951656E 951657E 9516571 9516572 9516575
10837-830EA 921.34 USD
10837-830 10837-832 10837-834 10837-836 10837-838 10837-840 10837-842 10837-844 10837-846 10837-848 10837-850 10837-852 10837-854 10837-856 10837-858 10838-908 10838-910 10838-912 10838-914 10838-916 10838-918 10838-920 10838-922 10838-924 10838-926 10838-944 10838-946 10838-948 10838-952 10838-954 10838-958 10838-960 10838-962 10838-964 10838-966 10838-968 10838-970 10838-972 10838-974 10840-110 10840-112 10840-114
Viva Biphenyl Columns, Restek
Chromatography Columns
Biphenyl offers a greater degree of dispersion than conventional phenyls and a greater degree of polarizability than phenyl hexyls, creating higher selectivity and a greater range of usability.

  • Rugged, spherical particles with 300 Å pore size
  • Particle size and pore size: 5 µm, spherical and 300 Å
  • Carbon load: 7%
  • End-cap: yes
  • pH range and temperature limit: 2.5 to 8 and 80 °C
  • USP phase: L11
  • Phase category and ligand type: phenyl and unique biphenyl

Because of these heightened interactions, this column shows substantial increases in retention—especially for dipolar, unsaturated, or conjugated solutes—and enhanced orthogonal selectivity when using methanol mobile phases. It is ideal for increasing sensitivity and selectivity in LC-MS analyses.

These have high proportion of pore/surface area available to large molecules and excellent for separating peptides or proteins.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR