Heaney-Ballentine Forceps, OR Grade, Sklar
Supplier: Sklar
90-2480 90-2580
10199-132EA 460.18 USD
10199-132 10199-134
Heaney-Ballentine Forceps, OR Grade, Sklar
Forceps
Sklar's Heaney-Ballentine Forceps are ratcheted, finger ring forceps used for grasping uterine ligaments during vaginal hysterectomy procedures
They are available straight or curved with a single toothed tip. Heaney-Ballentine forceps have longitudinal serrations which distinguish them from standard Heaney forceps, which have diagonal serrations.