Heaney Forceps, OR Grade, Sklar
Supplier: Sklar
90-1977 90-2177 90-2280 90-2387
10197-178EA 400.76 USD
10197-178 10199-088 10199-102 10199-130
Heaney Forceps, OR Grade, Sklar
Forceps
Sklar's Heaney Forceps are ratcheted, finger ring forceps used for grasping uterine ligaments during vaginal hysterectomy procedures
They are available straight or curved with one or two teeth. Heaney forceps have diagonal serrations the full length of the jaws, and the jaws are approximately 1/3 the length of the shanks. The combination of serrations and teeth provide a secure hold on the ligaments.