Pomeroy Ear Syringe, OR Grade, Sklar
Supplier: Sklar
67-3404 67-3402 67-3418
10193-352EA 425.28 USD
10193-352 10194-310 10198-362
Pomeroy Ear Syringe, OR Grade, Sklar
Syringes Syringes, Medical
Sklar's Pomery Ear Syringe is a three finger ring, ENT instrument used for irrigation and cleaning of the ear canal
The syringe is filled with body temperature water or other irrigation solution which is then injected into the ear canal. The flow and force of the solution losens and flushes out cerumen and debris. Two different tips are available for this syringe. One is bulbous and does not enter the ear canal deeply, whereas the other is conical and can enter further into the canal. The size and shape of the patient's ear, along with user preference, will determine which type of tip is utilized.
Order Now
SPECIFICATIONS
Environmentally Preferable