Ward's® Preserved Freshwater Perch
Supplier: Avantor
470001-500
470000-476PK 1113.95 USD
470000-476 470000-552 470000-778 470000-780 470000-908 470001-496 470001-498 470001-500
Ward's® Preserved Freshwater Perch
Educational Materials Biology Educational Materials Ecology Learning Activities
Formalin preserved perch, shipped in nontoxic Wardsafe. Available in vacuum packs or pails
  • Vertebrate anatomy
  • Bony fish anatomy
  • High quality preservation
Yellow perch, Perca flavescens, are bony fishes of the class Osteichthyes. Perch make a great dissection for middles chool, high school and college students alike. Introduce topics like vertebrate anatomy and bony fish anatmony. Ideal for studying external anatomical features, internal organs and organ systems. Plain and single-injected specimens are available. Single injected specimens are injectedMore Product Information