Ward's® Advanced Electrophoresis Bundles
Supplier: Avantor
470218-346KT 1099.99 USD
470218-346 470218-348 470218-350
Ward's® Advanced Electrophoresis Bundles
Educational Materials Biology Educational Materials Genetics Learning Activities
The professional-grade equipment in the Advanced Electrophoresis Bundles comes in a variety of sizes for any lab or class.
  • Professional-grade equipment
  • Bundles offer a great value
  • Different size bundles for any lab or class size
  • One year manufacturer's warranty on all equipment
  • Kit includes DNA kits, variable volume pipettors, and detailed instructions
Gel electrophoresis is a great way to introduce students to biotechnology and reinforce key genetics concepts. These convenient bundles are designed to provide all of the professional grade materials needed to perform gel electrophoresis with students.
Choose the size of equipment bundle that you need based on your class size. The advanced systems offer an affordable solution with easy-to-use DNA kits, variable volume pipettors, and stacking power supplies. Medium and large sets also include more equipment for additional lab applications.
Ordering information: Small Advanced Bundle contains 2 gel boxes; 1 stackable power supply; 2 variable-volume (20P) micropipettes; 2 racks of tips; 1 kit: "Introduction to Gel Electrophoresis with DNA".
Medium Advanced Bundle contains 4 gel boxes; 2 stackable power supplies; 8 variable-volume (20P) micropipettes; 4 racks of tips; 2 kits: Introduction to Gel Electrophoresis with DNA"; White-light gel Illuminator; 1 Bag of 1.5mL Sterile Microtubes; 1 Ward's Mini-centrifuge.
Large Advanced Bundle contains 6 gel boxes; 3 stackable power supplies; 12 variable-volume (20P) micropipettes; 10 racks of tips; 3 kits: Introduction to Gel Electrophoresis with DNA"; 1 White-light gel Illuminator; 2 Bags of 1.5mL Sterile Microtubes; 2 Ward's Mini-centrifuges; 1 Digital Water Bath (5.5L).