Jackson Improved Tracheostomy Tube, 4-Piece Set, OR Grade, Sklar
Supplier: Sklar
78-2032 78-2025 78-2026 78-2027 78-2028 78-2030 78-2024 78-2033 78-2031 78-2029
10198-194EA 543.93 USD
10198-194 10194-050 10194-280 10198-196 10194-282 10198-198 10198-200 10198-202 10194-048 10198-192
Jackson Improved Tracheostomy Tube, 4-Piece Set, OR Grade, Sklar
Cannulae
Sklar's Jackson Improved Style Tracheostomy four piece set.
The Jackson improved style features a small nub locking mechanism on the faceplate that works with a rotating lock on the inner cannula to secure it in place. A portion of the rotating lock is also cross-serrated so it can be gripped firmly and securely removed.
The obturator and cannulas work together to ensure that patients can breathe properly following a tracheal incision. The adaptor is utilized when items such as additional tubing, resusitation, or nebulizing devices are required. The outer cannula is the part of the device which keeps the actual incision in the neck open.
The improved style inner cannula does not have "wings." It is removed from the outer cannula by simply grasping the rotating lock and unlocking, which can easily be done because it is not flush with the faceplate, as is the top of the inner cannula on the original style device.
The obturator is used to safely and gently place the outer cannula into the incision and into the trachea. The neck plate, which is attached to the outer cannula, is where the side slots are located for tying or strapping the tube around the patient's neck for stability. Once the outer cannula is secured, the obturator is removed and the inner cannula is inserted.
Delivery information: Consists of one obturator, an inner cannula, an outer cannula, and a second inner cannula with a 15 mm adaptor attached.