Nystatin 10 mg/mL in methanol, sterile filtered
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Fungicidin, Mycostatin
J67369-8EQ
J67369-XF
10814-962PK
190.46
USD
10814-962
10814-964
Nystatin 10 mg/mL in methanol, sterile filtered
Nystatin
Liquid
Formula:
C₄₇H₇₅NO₁₇ Storage Temperature: Refrigerator |
MDL Number:
MFCD25541651 CAS Number: 1400-61-9 EINECS: 215-749-0 UN: 1230 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...