Calcium-D-saccharate tetrahydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Calcium D-glucarate tetrahydrate

J66947-22 J66947-14
10815-922EA 123 USD
10815-922 10815-920
Calcium-D-saccharate tetrahydrate 98%
Calcium-D-saccharate tetrahydrate

D-Saccharate (D-glucaric acid) is used to study its potential chemopreventive properties and anti-oxidative properties at the level of peroxynitrite-induced damages.


Soluble in water( 0,43 g/l)


Caution: Moisture Sensitive. Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Formula: C₆H₁₆CaO₁₂
Storage Temperature: Ambient
MDL Number: MFCD00150704
CAS Number: 5793-89-5
EINECS: 227-334-1
Merck Index: 14,1700

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR