Sucralose 98%, powder

Supplier: Thermo Scientific Chemicals

Synonyms: Trichlorosucrose, (2R,3R,4R,5R,6R)-2-(((2R,3S,4S,5S)-2,5-Bis(chloromethyl)-3,4-dihydroxytetrahydrofuran-2-yl)oxy)-5-chloro-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4-diol, 1-(1,6-Dichloro-1,6-dideoxy-beta-D-fructofuranosyl)-4-chloro-4-deoxy-alpha-D-galactopyranoside

J66736-18 J66736-22 J66736-30
10814-812EA 75.3 USD
10814-812 10814-814 10814-816
Sucralose 98%, powder
Sucralose

A synthetic sweetener approximately 600 times sweeter than sucrose. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells


Purity: 98% CAS No.: 56038-13-2 Molecular Formula: C12H19Cl3O8 Formula Weight: 397.63 Physical Form: Powder Appearance: White Melting Point: 130° dec. Beilstein Reference: 3654410 Merck Reference: 14,8880 MDL No.: MFCD03648615


A synthetic sweetener approximately 600 times sweeter than sucrose. Activates T1R2/T1R3 sweet taste receptors on enteroendocrine cells

Formula: C₁₂H₁₉Cl₃O₈
MW: 397.63 g/mol
Melting Pt: 130 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03648615
CAS Number: 56038-13-2
EINECS: 259-952-2
Merck Index: 14,8880

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR