Lactose

Supplier: Thermo Scientific Chemicals

Synonyms: Milk sugar

J66376-A1 J66376-36
10814-600EA 153 USD
10814-600 10814-598
Lactose
Lactose
Formula: C₁₂H₂₂O₁₁
MW: 342.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151251
CAS Number: 63-42-3
EINECS: 200-559-2
Merck Index: 14,5343

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR