Ethidium bromide 98% Dry weight Electrophoresis

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

J66192-03 J66192-06
10814-494EA 33.1 USD
10814-494 10814-496
Ethidium bromide 98% Dry weight Electrophoresis
Ethidium bromide

Purity: 98% CAS No.: 1239-45-8 Molecular Formula: C21H20BrN3 Formula Weight: 394.32 Appearance: Red to purple Odor: Weak Beilstein Reference: 3642536 Merck Reference: 14,4731 UN No.: 2811 MDL No.: MFCD00011724

Formula: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Boiling Pt: 72
Melting Pt: ∼ 261 °C
Storage Temperature: Ambient
MDL Number: MFCD00011724
CAS Number: 1239-45-8
EINECS: 214-984-6
UN: 2811
ADR: 6.1 III
Merck Index: 14,4731

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR