4-Methylumbelliferyl phosphate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 4-Methylumbelliferyl phosphoric acid4-Methyl-2-oxo-2H-chromen-7-yl dihydrogen phosphatePhosphoric Acid 4-Methylumbelliferyl Ester

J66091-MD J66091-03 J66091-06
10814-428EA 528.44 USD
10814-428 10814-426 10814-430
4-Methylumbelliferyl phosphate 99%
4-Methylumbelliferyl phosphate
Formula: C₁₀H₉O₆P
MW: 256.15 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Freezer
MDL Number: MFCD00016969
CAS Number: 3368-04-5
EINECS: 222-137-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR