Tricine 99% for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-(Tris(hydroxymethyl)methyl)glycine
J64437-36
J64437-22
10814-328EA
513.28
USD
10814-328
10814-326
Tricine 99% for electrophoresis
Tricine
Formula:
(HOCH₂)₃CNHCH₂CO₂H MW: 179.17 g/mol Melting Pt: ca 183 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004277 CAS Number: 5704-04-1 EINECS: 227-193-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...