4-Hexyloxyaniline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H55868-06
H55868-14
AAH55868-03EA
30.12
USD
AAH55868-03
AAH55868-06
AAH55868-14
4-Hexyloxyaniline 99%
4-Hexyloxyaniline
Purity: 99%
CAS No.: 39905-57-2
Molecular Formula: C12H19NO
Formula Weight: 193.29
Flash Point: 113°(235°F)
Physical Form: Solid
Melting Point: 43-45°
Boiling Point: 155-158°/5mm
MDL No.: MFCD00007868
Formula:
C₁₂H₁₉NO Boiling Pt: 155…158 °C/5 mm Melting Pt: 43…45 °C Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007868 CAS Number: 39905-57-2 EINECS: 254-696-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...