, pellets, Diameter 1-2mm (0.04-0.08in)
Supplier: Thermo Scientific Chemicals
Synonyms:
Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate
087957-A7
087957-A1
AA87957-A7EA
340.99
USD
AA87957-A7
AA87957-A1
, pellets, Diameter 1-2mm (0.04-0.08in)
Molecular sieve 3A (0.3 nm, 3 Å)
CAS No.: 308080-99-1
Physical Form: Pellets
Sensitivity: Hygroscopic
Appearance: Pale brown
MDL No.: MFCD00147627
Formula:
KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O Storage Temperature: Ambient |
MDL Number:
MFCD00131613 CAS Number: 308080-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...