Glassy carbon, spherical powder 200-400 µm Type2

Supplier: Thermo Scientific Chemicals
Warning

042550-18 042550-09 042550-30
AA42550-18EA 131 USD
AA42550-18 AA42550-09 AA42550-30
Glassy carbon, spherical powder 200-400 µm Type2
Glassy carbon

CAS No.: 7440-44-0 Odor: Odorless UN No.: 1325 MDL No.: MFCD00133992

Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR