Graphite Electrode, Crater, Thermo Scientific
Supplier: Thermo Scientific Chemicals
014743-KN 040788-KN
AA14743-KNEA 516 USD
AA14743-KN AA40788-KN
Graphite Electrode, Crater, Thermo Scientific
Electrodes Other Electrodes
Graphite electrodes are used mainly in electric arc furnace steel production
  • AGKSP grade
  • Ultra F purity
  • Insoluble in water
It also used to refine steel in ladle furnaces and in other smelting processes, furnance design, sharp requirements, water spray rings and charging practice.