Glassy carbon, powder 80-200 µm Type 1

Supplier: Thermo Scientific Chemicals
Warning

038002-09 038002-18
AA38002-09EA 120 USD
AA38002-09 AA38002-18
Glassy carbon, powder 80-200 µm Type 1
Glassy carbon

UN No.: 1325 MDL No.: MFCD00133992

Formula: C
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR