Graphite (natural) ≥99.9995% (metals basis), powder 2-15 µm, microcrystal grade

Supplier: Thermo Scientific Chemicals
Warning

014736-TB 014736-TC 014736-TA 014736-TD
AA14736-TBEA 211 USD
AA14736-TB AA14736-TC AA14736-TA AA14736-TD
Graphite (natural) ≥99.9995% (metals basis), powder 2-15 µm, microcrystal grade
Graphite

Purity: 99.9995% Physical Form: UCP-1-M grade, Ultra "F" purity Appearance: Black Odor: Odorless UN No.: 1325 MDL No.: MFCD00144065

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
Storage Temperature: Ambient
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
UN: 1325
ADR: 4.1 III
Merck Index: 12,04560

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR