Sklar Black™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Supplier: Sklar
90-9106 90-9109 90-9100 90-9102 90-9103
10184-234EA 1246.34 USD
10184-234 10184-286 10184-292 10184-294 10184-296
Sklar Black™ Auvard Weighted Vaginal Speculum, OR Grade, Sklar
Specula
The Sklar Black™ Auvard Weighted Vaginal Speculum is a single end vaginal speculum commonly used in laser OB/GYN procedures.
It is available with various blades styles and three weight choices in order to accommodate the needs of the patient and the procedure. The weight holds the speculum in place and provides an open view for the surgeon to perform procedures.
Sklar Black™ Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less thanMore Product Information