Econo™ Sterile Pederson Vaginal Speculum, Floor Grade, Sklar
Supplier: Sklar
96-2612 96-2613 96-2614 96-2615 96-2616 96-2617
10184-662CS 2219.15 USD
10184-662 10184-664 10184-666 10184-668 10184-670 10184-672
Econo™ Sterile Pederson Vaginal Speculum, Floor Grade, Sklar
Specula
The Econo™ Sterile Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix.
It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are more narrow than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination. They are available in bulk quantities.