Anti-RAF1 Rabbit Polyclonal Antibody

Supplier: R & D Systems
PPS061
220022-171EA 663.78 USD
220022-171
Anti-RAF1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Dog, Chicken, Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR