Anti-CK18 Mouse Monoclonal Antibody [clone: 810811]

Supplier: R & D Systems
MAB7619
102887-266EA 620.4 USD
102887-266
Anti-CK18 Mouse Monoclonal Antibody [clone: 810811]
Antibodies
Simple Western

Type: Primary
Antigen: CK18
Clonality: Monoclonal
Clone: 810811
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR