Anti-IL4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3007]

Supplier: R&D Systems
IC204P-025 IC204P-100
102884-418EA 287.78 USD
102884-418 102884-420
Anti-IL4 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3007]
Antibodies
Type: Primary
Antigen: IL4
Clonality: Monoclonal
Clone: 3007
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR