Anti-CXCR6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 56811]

Supplier: R & D Systems
FAB699A-100 FAB699A-025
102883-050EA 229.94 USD
102883-050 102883-052
Anti-CXCR6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 56811]
Antibodies
Type: Primary
Antigen: CXCR6
Clonality: Monoclonal
Clone: 56811
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR