Anti-CMKLR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 84939]

Supplier: R & D Systems
FAB362P
102882-348EA 562.55 USD
102882-348
Anti-CMKLR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 84939]
Antibodies
Type: Primary
Antigen: CMKLR1
Clonality: Monoclonal
Clone: 84939
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR