Anti-WIF1 Mouse Monoclonal Antibody (Biotin) [clone: 133016]

Supplier: R & D Systems
BAM1341
220014-555EA 765.02 USD
220014-555
Anti-WIF1 Mouse Monoclonal Antibody (Biotin) [clone: 133016]
Antibodies
Type: Primary
Antigen: WIF1
Clonality: Monoclonal
Clone: 133016
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR