Anti-KLK11 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF1595
220013-085EA 721.63 USD
220013-085
Anti-KLK11 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: KLK11
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR