Anti-ANGPTL3 Goat Polyclonal Antibody (Biotin)

Supplier: R & D Systems
BAF136
220012-984EA 779.48 USD
220012-984
Anti-ANGPTL3 Goat Polyclonal Antibody (Biotin)
Antibodies
Type: Primary
Antigen: ANGPTL3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR