Anti-TCN2 Sheep Polyclonal Antibody

Supplier: R & D Systems
AF7895
102878-852EA 663.78 USD
102878-852
Anti-TCN2 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TCN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR