Anti-EDAR Goat Polyclonal Antibody

Supplier: R & D Systems
AF745
220012-537EA 663.78 USD
220012-537
Anti-EDAR Goat Polyclonal Antibody
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: EDAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR