Anti-IRAK3 Sheep Polyclonal Antibody

Supplier: R & D Systems
AF6264
220012-452EA 663.78 USD
220012-452
Anti-IRAK3 Sheep Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IRAK3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR