Anti-SIRPA Sheep Polyclonal Antibody

Supplier: R & D Systems
AF4546
220011-690EA 663.78 USD
220011-690
Anti-SIRPA Sheep Polyclonal Antibody
Antibodies
Simple Western

Type: Primary
Antigen: SIRPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR