Anti-CNPY3 Goat Polyclonal Antibody

Supplier: R & D Systems
AF4429
220011-634EA 663.78 USD
220011-634
Anti-CNPY3 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CNPY3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR