Anti-CSF1 Goat Polyclonal Antibody

Supplier: R & D Systems
AF416
220011-502EA 707.17 USD
220011-502
Anti-CSF1 Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CSF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR